APO455834905 Apollo Scientific Ltd

3-Chloro-4-fluorobenzylamine

  • MDL number MFCD00042507
  • Cas Number : 72235-56-4
  • Molecular Weight : 159.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 104-106/15mm
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzylamine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart