APO455834909 Apollo Scientific Ltd

2-Chloro-6-fluorobenzylthiol

  • MDL number MFCD00052473
  • Cas Number : 170924-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-fluorobenzyl mercaptan, (2-Chloro-6-fluorophenyl)methanethiol
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 102/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=837

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=837
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart