APO455834913 Apollo Scientific Ltd

2-Chloro-4-fluoroiodobenzene

  • MDL number MFCD00051774
  • Cas Number : 101335-11-9
  • Molecular Weight : 256.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-4-fluoro-1-iodobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 96-98/15mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluoroiodobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluoroiodobenzene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart