APO455834919 Apollo Scientific Ltd

2-Hydroxy-6-(trifluoromethyl)nicotinic acid

  • MDL number MFCD01862657
  • Cas Number : 191595-63-8
  • Molecular Weight : 207.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-167
Synonym 2-Hydroxy-6-(trifluoromethyl)pyridine-3-carboxylic acid, 3-Carboxy-2-hydroxy-6-(trifluoromethyl)pyridine
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxy-6-trifluoromethylnicotinic-acid

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/2-hydroxy-6-trifluoromethylnicotinic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart