APO455834921 Apollo Scientific Ltd

2-Chloro-5-fluorophenol

  • MDL number MFCD00042524
  • Cas Number : 3827-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 183-185
Product Link http://www.apolloscientific.co.uk/display_item.php?id=859

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=859
pictograms GHS05;GHS07;GHS09
hstatements H302;H312;H314;H411
Availability details for
Quantity
  • Availability
  • Add to Cart