APO455834928 Apollo Scientific Ltd

3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonyl chloride

  • MDL number MFCD00055650
  • Cas Number : 69399-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-52
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=878

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=878
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart