APO455834929 Apollo Scientific Ltd

2-Chloro-4-fluorothiophenol

  • MDL number MFCD00052936
  • Cas Number : 175277-99-3
  • Molecular Weight : 162.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-fluorobenzenethiol
Inchi Code 1S/C6H4ClFS/c7-5-3-4(8)1-2-6(5)9/h1-3,9H
Inchi Key KUAPPJSILOMQPC-UHFFFAOYSA-N

Description

Synonym 2-Chloro-4-fluorobenzenethiol, 2-Chloro-4-fluorophenyl mercaptan
Purity 97%
Flash Point 86-88/15mm
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluorothiophenol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluorothiophenol
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart