APO455834931 Apollo Scientific Ltd

2-Chloro-5-fluorotoluene

  • MDL number MFCD00040853
  • Cas Number : 33406-96-1
  • Molecular Weight : 144.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-4-fluoro-2-methylbenzene
Density 1.2
Flash Point 51
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 156-157
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-fluorotoluene

Safety Information

signalword Warning
pcodes P280;P210
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-fluorotoluene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart