APO455834933 Apollo Scientific Ltd

3-Chloro-2-fluoropyridine

  • MDL number MFCD04114144
  • Cas Number : 1480-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.33
Flash Point 52
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 162
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-fluoropyridine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-fluoropyridine
pictograms GHS02;GHS05;GHS07
hstatements H226;H314
Availability details for
Quantity
  • Availability
  • Add to Cart