APO455834948 Apollo Scientific Ltd

1-Chloro-2-iodotetrafluoroethane

  • MDL number MFCD00069092
  • Cas Number : 421-78-3
  • Molecular Weight : 262.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chlorotetrafluoroethyl iodide
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 56
Product Link https://store.apolloscientific.co.uk/product/1-chloro-2-iodotetrafluoroethane

Safety Information

signalword Danger
pcodes P260;P280;P301+P310
MSDS https://store.apolloscientific.co.uk/product/1-chloro-2-iodotetrafluoroethane
pictograms GHS06;GHS08
hstatements H301+H311+H331;H351
Availability details for
Quantity
  • Availability
  • Add to Cart