APO455834961 Apollo Scientific Ltd

3-Fluoro-2-hydrazinopyridine

  • MDL number MFCD09998071
  • Cas Number : 887266-57-1
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Fluoropyridin-2-yl)hydrazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-hydrazinopyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-hydrazinopyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart