APO455834963 Apollo Scientific Ltd

1-Acetyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD01320690
  • Cas Number : 1017793-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[3-Methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazol-1-yl]ethan-1-one, 1-Ethanoyl-3-methyl-4-nitro-5-(trifluoromethyl)-1H-pyrazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10037

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10037
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart