APO455834965 Apollo Scientific Ltd

1,3-Benzodioxol-5-yl-4,4,4-trifluorobutane-1,3-dione

  • MDL number MFCD01570563
  • Cas Number : 306935-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-108
Synonym 3-[3,4-(Methylenedioxy)benzoyl]-1,1,1-trifluoroacetone
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3919

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3919
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart