APO455834966 Apollo Scientific Ltd

3-Amino-5-chloro-4-hydroxybenzotrifluoride

  • MDL number MFCD07777110
  • Cas Number : 78068-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-6-chloro-4-(trifluoromethyl)phenol, 3-Chloro-2-hydroxy-5-(trifluoromethyl)aniline
Purity 98
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10050

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10050
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart