APO455834967 Apollo Scientific Ltd

2-Amino-5-fluorobenzamide

  • MDL number MFCD03428522
  • Cas Number : 63069-49-8
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-148
Synonym 2-Carbamoyl-4-fluoroaniline, 5-Fluoroanthranilamide
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-fluorobenzamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-fluorobenzamide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart