APO455834973 Apollo Scientific Ltd

3,5-Difluoro-alpha-methylbenzylamine

  • MDL number MFCD04038286
  • Cas Number : 321318-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3,5-Difluorophenyl)ethylamine
Purity 97
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 175
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-alpha-methylbenzylamine

Safety Information

signalword Danger
pcodes P280;P301+P312;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-alpha-methylbenzylamine
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart