APO455834976 Apollo Scientific Ltd

4-Chloro-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD00190117
  • Cas Number : 1735-54-2
  • Molecular Weight : 205.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-68
Synonym 2-Chloro-5-cyanobenzotrifluoride, 4-Chloro-alpha,alpha,alpha-trifluoro-m-tolunitrile
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 100-105/11mm
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-trifluoromethylbenzonitrile

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-trifluoromethylbenzonitrile
pictograms GHS06
hstatements H301+H311+H331;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart