APO455834979 Apollo Scientific Ltd

2-Amino-5-iodobenzotrifluoride

  • MDL number MFCD04972657
  • Cas Number : 97760-97-9
  • Molecular Weight : 287.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Iodo-2-(trifluoromethyl)aniline, 4-Iodo-alpha,alpha,alpha-trifluoro-o-toluidine
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-iodobenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-iodobenzotrifluoride
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart