APO455834982 Apollo Scientific Ltd

1-(4-Chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride

  • MDL number MFCD00068143
  • Cas Number : 175137-19-6
  • Molecular Weight : 309.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Synonym 2-(4-Chlorophenyl)-3-(trifluoromethyl)-2H-pyrazole-4-carbonyl chloride
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 120/0.3mm
Product Link https://store.apolloscientific.co.uk/product/1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carbonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carbonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart