APO455834984 Apollo Scientific Ltd

3-Chlorotetrafluoropropanoic acid

  • MDL number MFCD00153099
  • Cas Number : 661-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.63
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 130-131
Product Link https://store.apolloscientific.co.uk/product/3-chlorotetrafluoropropanoic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chlorotetrafluoropropanoic-acid
pictograms GHS05
hstatements H290;H314
Availability details for
Quantity
  • Availability
  • Add to Cart