APO455834995 Apollo Scientific Ltd

5-Methoxy-4'-(trifluoromethyl)valerophenone

  • MDL number MFCD06658173
  • Cas Number : 61718-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Synonym 5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one, 4-(5-Methoxypentanoyl)benzotrifluoride
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-methoxy-4-trifluoromethylvalerophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methoxy-4-trifluoromethylvalerophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart