APO455834997 Apollo Scientific Ltd

2-Chloro-1,1,2-trifluoroethyl ethyl ether

  • MDL number MFCD00018849
  • Cas Number : 310-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.273
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 82/630mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=979

Safety Information

signalword Danger
pcodes P210;P241;P280;P303+P361+P353;P304+P340;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=979
pictograms GHS02;GHS07
hstatements H225;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart