APO455835009 Apollo Scientific Ltd

3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine

  • MDL number MFCD00067865
  • Cas Number : 89570-82-1
  • Molecular Weight : 211.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-94
Synonym [3-Chloro-5-(trifluoromethyl)pyridin-2-yl]hydrazine, 5-Chloro-6-hydrazino-alpha,alpha,alpha-trifluoro-3-picoline
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-hydrazino-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-hydrazino-5-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart