APO455835012 Apollo Scientific Ltd

1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperazine

  • MDL number MFCD00173934
  • Cas Number : 132834-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-67
Synonym 3-Chloro-2-(piperazin-1-yl)-5-(trifluoromethyl)pyridine, 5-Chloro-6-(piperazin-1-yl)-alpha,alpha,alpha-trifluoro-3-picoline
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1021

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1021
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart