APO455835013 Apollo Scientific Ltd

2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride

  • MDL number MFCD00173789
  • Cas Number : 154934-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-5-(chlorocarbonyl)-4-(trifluoromethyl)pyrimidine, 2-Chloro-5-(chloroformyl)-4-(trifluoromethyl)-1,3-diazine
Purity 97
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 54/0.15mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-trifluoromethylpyrimidine-5-carbonyl-chloride

Safety Information

signalword Danger
pcodes P232;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-trifluoromethylpyrimidine-5-carbonyl-chloride
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart