APO455835016 Apollo Scientific Ltd

(1H,1H,2H,2H-Heptadecafluorodec-1-yl)(trimethoxy)silane

  • MDL number MFCD07368748
  • Cas Number : 83048-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodec-1-yl)(trimethoxy)silane, 1H,1H,2H,2H-Perfluorodecyltrimethoxysilane
Purity 97
Density 1.54
Flash Point >65
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 83-84/1mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8932

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8932
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart