APO455835019 Apollo Scientific Ltd

2,6-Dichloro-4-(trifluoromethyl)nicotinonitrile

  • MDL number MFCD00084977
  • Cas Number : 13600-42-5
  • Molecular Weight : 241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-40
Synonym 3-Cyano-2,6-dichloro-4-(trifluoromethyl)pyridine, 2,6-Dichloro-4-(trifluoromethyl)pyridine-3-carbonitrile
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-4-trifluoromethylnicotinonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-4-trifluoromethylnicotinonitrile
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart