APO455835022 Apollo Scientific Ltd

4-Fluoro-1-naphthonitrile

  • MDL number MFCD00236656
  • Cas Number : 13916-99-9
  • Molecular Weight : 171.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Synonym 4-Fluoronaphthalene-1-carbonitrile, 1-Cyano-4-fluoronaphthalene
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-1-naphthonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-1-naphthonitrile
pictograms GHS07
hstatements H302;H312;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart