APO455835024 Apollo Scientific Ltd

4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzonitrile

  • MDL number MFCD00173780
  • Cas Number : 175277-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Synonym 2-(4-Cyanophenoxy)-5-(trifluoromethyl)pyridine
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1055

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1055
pictograms GHS07
hstatements H302;H312;H332
Availability details for
Quantity
  • Availability
  • Add to Cart