APO455835029 Apollo Scientific Ltd

5-Methyl-4-nitro-3-(trifluoromethyl)pyrazole

  • MDL number MFCD00156055
  • Cas Number : 27116-80-9
  • Molecular Weight : 195.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 74-79
Iupac Name 5-methyl-4-nitro-3-(trifluoromethyl)-1H-pyrazole
Inchi Code 1S/C5H4F3N3O2/c1-2-3(11(12)13)4(10-9-2)5(6,7)8/h1H3,(H,9,10)
Inchi Key MZVQCTATYLWIQA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 74-79
Product Link https://store.apolloscientific.co.uk/product/5-methyl-4-nitro-3-trifluoromethylpyrazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-4-nitro-3-trifluoromethylpyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart