APO455835036 Apollo Scientific Ltd

4-(Benzyloxy)benzotrifluoride

  • MDL number MFCD00275905
  • Cas Number : 70097-65-3
  • Molecular Weight : 252.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Benzyloxy-4-(trifluoromethyl)benzene, Benzyl 4-(trifluoromethyl)phenyl ether
Purity 98
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-benzyloxybenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-benzyloxybenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart