APO455835042 Apollo Scientific Ltd

2-Amino-2-methyl-3,3,3-trifluoropropanoic acid, N-CBZ protected

  • MDL number MFCD07777230
  • Cas Number : 139520-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha-(Trifluoromethyl)-DL-alanine, N-CBZ protected, Benzyl (2-carboxy-1,1,1-trifluoroprop-2-yl)carbamate, 2-{[(Benzyloxy)carbonyl]amino}-2-methyl-3,3,3-trifluoropropanoic acid
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10085

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10085
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart