APO455835046 Apollo Scientific Ltd

2,4-Diamino-6-(4-fluorophenyl)pyrimidine

  • MDL number MFCD00052076
  • Cas Number : 175137-25-4
  • Molecular Weight : 204.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-166
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-diamino-6-4-fluorophenylpyrimidine

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/24-diamino-6-4-fluorophenylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart