APO455835056 Apollo Scientific Ltd

1,3-Bis(1H,1H,2H,2H-tridecafluoroooct-1-yl)-1,1,3,3-tetramethyldisiloxane

  • MDL number MFCD00270970
  • Cas Number : 71363-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,3-Bis(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-yl)-1,1,3,3-tetramethyldisiloxane
Density 1.46
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 81-82/11mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10142

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10142
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart