APO455835074 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)-2-bromoaniline

  • MDL number MFCD00134419
  • Cas Number : 174824-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-37
Synonym 5-Amino-4-bromo-alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-m-xylene
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10178

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10178
pictograms GHS06
hstatements H311;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart