APO455835075 Apollo Scientific Ltd

3-Bromo-2-chlorobenzotrifluoride

  • MDL number MFCD04115994
  • Cas Number : 56131-47-6
  • Molecular Weight : 259.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-chloro-3-(trifluoromethyl)benzene
Inchi Code 1S/C7H3BrClF3/c8-5-3-1-2-4(6(5)9)7(10,11)12/h1-3H
Inchi Key DWTJMEQXGJMZMM-UHFFFAOYSA-N

Description

Synonym 1-Bromo-2-chloro-3-(trifluoromethyl)benzene
Purity 98%
Flash Point 75-77/0.5mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-chlorobenzotrifluoride

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-chlorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart