APO455835082 Apollo Scientific Ltd

4,5-Difluoro-1H-benzimidazole

  • MDL number MFCD00153122
  • Cas Number : 236736-21-3
  • Molecular Weight : 154.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211-213
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-difluoro-1h-benzimidazole

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-difluoro-1h-benzimidazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart