APO455835084 Apollo Scientific Ltd

2-[1-Methyl-3-(trifluoromethyl)pyrazol-5-yl]thiophene-5-carboxaldehyde

  • MDL number MFCD00052158
  • Cas Number : 175202-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-78
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4081

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4081
pictograms GHS06;GHS07
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart