APO455835087 Apollo Scientific Ltd

2,6-Dibromo-4-(trifluoromethoxy)aniline

  • MDL number MFCD00153113
  • Cas Number : 88149-49-9
  • Molecular Weight : 334.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-72
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 65/0.1mm
Product Link https://store.apolloscientific.co.uk/product/26-dibromo-4-trifluoromethoxyaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dibromo-4-trifluoromethoxyaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart