APO455835088 Apollo Scientific Ltd

2-Amino-3,5-dibromobenzotrifluoride

  • MDL number MFCD00178769
  • Cas Number : 71757-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-43
Synonym 2,4-Dibromo-6-(trifluoromethyl)aniline, 4,6-Dibromo-alpha,alpha,alpha-trifluoro-o-toluidine
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 57-59/0.7mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1129

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1129
pictograms GHS06
hstatements H302;H332;H311;H335
Availability details for
Quantity
  • Availability
  • Add to Cart