APO455835090 Apollo Scientific Ltd

1,2-Dibromo-3,3,3-trifluoroprop-1-ene

  • MDL number MFCD00042236
  • Cas Number : 431-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 96
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1136

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1136
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart