APO455835114 Apollo Scientific Ltd

2,3-Dichlorofluorobenzene

  • MDL number MFCD00060647
  • Cas Number : 36556-50-0
  • Molecular Weight : 164.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2-Dichloro-3-fluorobenzene
Purity 99
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 80/22mm
Product Link https://store.apolloscientific.co.uk/product/23-dichlorofluorobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-dichlorofluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart