APO455835115 Apollo Scientific Ltd

2,4-Dichlorofluorobenzene

  • MDL number MFCD00000580
  • Cas Number : 1435-48-9
  • Molecular Weight : 164.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 37
bp -23
Iupac Name 2,4-dichloro-1-fluorobenzene
density 1.409
Inchi Code 1S/C6H3Cl2F/c7-4-1-2-6(9)5(8)3-4/h1-3H
Inchi Key BDJZCCWUSOZUQG-UHFFFAOYSA-N

Description

Melting Point 37
Synonym 2,4-Dichloro-1-fluorobenzene
Purity 99%
Density 1.409
Flash Point 172-174
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point -23
Product Link https://store.apolloscientific.co.uk/product/24-dichlorofluorobenzene

Safety Information

signalword Warning
pcodes P280;P271;P260
MSDS https://store.apolloscientific.co.uk/product/24-dichlorofluorobenzene
pictograms GHS02;GHS07;GHS08;GHS09
hstatements H226;H302;H315;H411
Availability details for
Quantity
  • Availability
  • Add to Cart