APO455835118 Apollo Scientific Ltd

3,4-Dichlorofluorobenzene

  • MDL number MFCD00018119
  • Cas Number : 1435-49-0
  • Molecular Weight : 164.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -1
Synonym 1,2-Dichloro-4-fluorobenzene
Purity 99
Density 1.40
Flash Point 64
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 166-171
Product Link https://store.apolloscientific.co.uk/product/34-dichlorofluorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/34-dichlorofluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart