APO455835127 Apollo Scientific Ltd

2-Methyl-5-(trifluoromethyl)benzyl chloride

  • MDL number MFCD01861155
  • Cas Number : 225656-63-3
  • Molecular Weight : 208.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Chloromethyl)-4-methylbenzotrifluoride, 2-(Chloromethyl)-4-(trifluoromethyl)toluene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-5-trifluoromethylbenzyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-5-trifluoromethylbenzyl-chloride
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart