APO455835133 Apollo Scientific Ltd

{[4-(Trifluoromethyl)phenyl]thio}acetic acid

  • MDL number MFCD00135062
  • Cas Number : 102582-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-118
Synonym {[4-(Trifluoromethyl)phenyl]sulphanyl}acetic acid, 4-[(Carboxymethyl)thio]benzotrifluoride
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=11422

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=11422
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart