APO455835151 Apollo Scientific Ltd

4'-(tert-Butyl)-2,2,2-trifluoroacetophenone

  • MDL number MFCD01319979
  • Cas Number : 73471-97-3
  • Molecular Weight : 230.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[4-(tert-Butyl)phenyl]-2,2,2-trifluoroethan-1-one
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butyl-222-trifluoroacetophenone

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-tert-butyl-222-trifluoroacetophenone
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart