APO455835156 Apollo Scientific Ltd

5-Bromo-2-fluoroiodobenzene

  • MDL number MFCD07368751
  • Cas Number : 116272-41-4
  • Molecular Weight : 300.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 103
Iupac Name 4-bromo-1-fluoro-2-iodobenzene
density 2.281
Inchi Code 1S/C6H3BrFI/c7-4-1-2-5(8)6(9)3-4/h1-3H
Inchi Key JNETZJWWXCLUKM-UHFFFAOYSA-N

Description

Melting Point 103
Synonym 4-Bromo-1-fluoro-2-iodobenzene
Purity 99%
Density 2.281
Flash Point 247
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-fluoroiodobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-fluoroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart