APO455835169 Apollo Scientific Ltd

alpha,alpha,beta,beta,4-Pentafluorophenetole

  • MDL number MFCD08059513
  • Cas Number : 887268-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Fluoro-4-(1,1,2,2-tetrafluoroethoxy)benzene, 4-Fluorophenyl 1,1,2,2-tetrafluoroethyl ether
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10331

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10331
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart