APO455835173 Apollo Scientific Ltd

2',3'-Difluoroacetophenone

  • MDL number MFCD00061274
  • Cas Number : 18355-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2,3-Difluorophenyl)ethan-1-one
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 85/20mm
Product Link https://store.apolloscientific.co.uk/product/23-difluoroacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoroacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart